Assay Details
| Gene Symbol: Washc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837968.3 | XP_004838025.1 | 25 - 26 | 3368 | 59 | |
| XM_021252142.1 | XP_021107801.1 | 25 - 26 | 3368 | 59 | ||
| XM_021252144.1 | XP_021107803.1 | 23 - 24 | 3125 | 59 | ||
| XM_021252153.1 | XP_021107812.1 | 22 - 23 | 2801 | 59 | ||
More Information
Set Membership: |
|

Washc5
Genomic Map