Assay Details
| Gene Symbol: Mia2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078883.2 | XP_012934337.2 | 25 - 26 | 3871 | 58 | |
| XM_021242838.1 | XP_021098497.1 | 19 - 20 | 1917 | 58 | ||
| XM_021242839.1 | XP_021098498.1 | 18 - 19 | 1788 | 58 | ||
More Information
Set Membership: |
|

Mia2
Genomic Map