Assay Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853788.2 | XP_004853845.1 | 3 - 4 | 327 | 64 | |
| XM_013075610.2 | XP_012931064.1 | 3 - 4 | 327 | 64 | ||
More Information
Set Membership: |
|

Tspan2
Genomic Map