Assay Details
| Gene Symbol: Hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852803.2 | XP_004852860.1 | 3 - 4 | 1007 | 70 | |
| XM_004852805.2 | XP_004852862.1 | 4 - 5 | 640 | 70 | ||
| XM_004852806.2 | XP_004852863.1 | 4 - 5 | 623 | 70 | ||
| XM_021264832.1 | XP_021120491.1 | 4 - 5 | 763 | 70 | ||
More Information
Set Membership: |
|

Hddc3
Genomic Map