Assay Details
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004861561.3 | XP_004861618.1 | 7 - 8 | 2418 | 53 | |
XM_004861562.3 | XP_004861619.1 | 6 - 7 | 2329 | 53 | ||
XM_021241678.1 | XP_021097337.1 | 7 - 8 | 2426 | 53 | ||
XM_021241679.1 | XP_021097338.1 | 6 - 7 | 2231 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
