Assay Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856860.2 | XP_004856917.1 | 2 - 3 | 838 | 94 | |
| XM_021237920.1 | XP_021093579.1 | 3 - 4 | 977 | 94 | ||
| XM_021237921.1 | XP_021093580.1 | 4 - 5 | 1120 | 94 | ||
| XM_021237922.1 | XP_021093581.1 | 3 - 4 | 937 | 94 | ||
More Information
Set Membership: |
|

Fbxo5
Genomic Map