Assay Details
| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854247.2 | XP_004854304.1 | 1 - 2 | 414 | 73 | |
| XM_004854248.2 | XP_004854305.1 | 2 - 3 | 320 | 73 | ||
| XM_004854249.2 | XP_004854306.1 | 2 - 3 | 366 | 73 | ||
| XM_004854251.2 | XP_004854308.1 | 1 - 2 | 414 | 73 | ||
| XM_004854252.2 | XP_004854309.1 | 1 - 2 | 414 | 73 | ||
| XM_013075657.2 | XP_012931111.1 | 1 - 2 | 415 | 73 | ||
| XM_021266080.1 | XP_021121739.1 | 1 - 2 | 415 | 73 | ||
More Information
Set Membership: |
|

Slc22a13
Genomic Map