Assay Details
| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854247.2 | XP_004854304.1 | 3 - 4 | 687 | 94 | |
| XM_004854248.2 | XP_004854305.1 | 4 - 5 | 593 | 94 | ||
| XM_004854249.2 | XP_004854306.1 | 4 - 5 | 639 | 94 | ||
| XM_004854251.2 | XP_004854308.1 | 3 - 4 | 687 | 94 | ||
| XM_004854252.2 | XP_004854309.1 | 3 - 4 | 687 | 94 | ||
| XM_013075657.2 | XP_012931111.1 | 3 - 4 | 688 | 94 | ||
| XM_021266080.1 | XP_021121739.1 | 3 - 4 | 688 | 94 | ||
More Information
Set Membership: |
|

Slc22a13
Genomic Map