Assay Details
| Gene Symbol: Cfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852374.3 | XP_004852431.1 | 1 - 2 | 208 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004852374.3 transcripts.
Set Membership: |
|

Cfl1
Genomic Map