Assay Details
| Gene Symbol: Cfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852374.3 | XP_004852431.1 | 3 - 4 | 591 | 68 | |
| XM_021264574.1 | XP_021120233.1 | 3 - 4 | 576 | 68 | ||
More Information
Set Membership: |
|

Cfl1
Genomic Map