Assay Details
| Gene Symbol: Znf532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858413.3 | XP_004858470.1 | 2 - 3 | 793 | 108 | |
| XM_004858414.3 | XP_004858471.1 | 3 - 4 | 395 | 108 | ||
| XM_004858415.3 | XP_004858472.1 | 2 - 3 | 313 | 108 | ||
| XM_021239292.1 | XP_021094951.1 | 3 - 4 | 875 | 108 | ||
| XM_021239293.1 | XP_021094952.1 | 2 - 3 | 308 | 108 | ||
| XM_021239294.1 | XP_021094953.1 | 2 - 3 | 334 | 108 | ||
| XM_021239295.1 | XP_021094954.1 | 2 - 3 | 1194 | 108 | ||
| XM_021239296.1 | XP_021094955.1 | 2 - 3 | 1190 | 108 | ||
| XM_021239297.1 | XP_021094956.1 | 2 - 3 | 1195 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004858413.3, XM_004858414.3, XM_004858415.3, XM_021239292.1, XM_021239293.1, XM_021239294.1, XM_021239295.1, XM_021239296.1, XM_021239297.1 transcripts.
Set Membership: |
|

Znf532
Genomic Map