Assay Details
| Gene Symbol: Znf532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858413.3 | XP_004858470.1 | 8 - 9 | 4066 | 76 | |
| XM_004858414.3 | XP_004858471.1 | 9 - 10 | 3668 | 76 | ||
| XM_004858415.3 | XP_004858472.1 | 8 - 9 | 3586 | 76 | ||
| XM_021239292.1 | XP_021094951.1 | 9 - 10 | 4148 | 76 | ||
| XM_021239293.1 | XP_021094952.1 | 8 - 9 | 3581 | 76 | ||
| XM_021239294.1 | XP_021094953.1 | 8 - 9 | 3607 | 76 | ||
| XM_021239295.1 | XP_021094954.1 | 5 - 6 | 3945 | 76 | ||
| XM_021239296.1 | XP_021094955.1 | 8 - 9 | 3875 | 76 | ||
| XM_021239297.1 | XP_021094956.1 | 4 - 5 | 3664 | 76 | ||
More Information
Set Membership: |
|

Znf532
Genomic Map