Assay Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835933.2 | XP_004835990.1 | 1 - 2 | 702 | 106 | |
| XM_004835934.2 | XP_004835991.1 | 1 - 2 | 1204 | 106 | ||
| XM_021266152.1 | XP_021121811.1 | 1 - 2 | 1202 | 106 | ||
More Information
Set Membership: |
|

Kcnj3
Genomic Map