Assay Details
| Gene Symbol: Mrpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873700.3 | XP_004873757.1 | 4 - 5 | 739 | 76 | |
| XM_021251433.1 | XP_021107092.1 | 3 - 4 | 827 | 76 | ||
More Information
Set Membership: |
|

Mrpl28
Genomic Map