Assay Details
| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 1 - 2 | 378 | 84 | |
| XM_021248902.1 | XP_021104561.1 | 1 - 2 | 379 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH17orf75| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 1 - 2 | 378 | 84 | |
| XM_021248902.1 | XP_021104561.1 | 1 - 2 | 379 | 84 | ||