Assay Details
| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 3 - 4 | 584 | 106 | |
| XM_021248902.1 | XP_021104561.1 | 3 - 4 | 585 | 106 | ||
More Information
Set Membership: |
|

CUNH17orf75
Genomic Map