Assay Details
| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 5 - 6 | 784 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH17orf75| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 5 - 6 | 784 | 90 | |