Assay Details
| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 7 - 8 | 966 | 95 | |
| XM_021248902.1 | XP_021104561.1 | 7 - 8 | 874 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH17orf75| Gene Symbol: CUNH17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870594.3 | XP_004870651.1 | 7 - 8 | 966 | 95 | |
| XM_021248902.1 | XP_021104561.1 | 7 - 8 | 874 | 95 | ||