Assay Details
| Gene Symbol: Otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837051.2 | XP_004837108.1 | 3 - 4 | 444 | 57 | |
| XM_004837052.2 | XP_004837109.1 | 3 - 4 | 449 | 57 | ||
More Information
Set Membership: |
|

Otub2
Genomic Map