Assay Details
| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845509.3 | XP_004845566.1 | 1 - 2 | 326 | 69 | |
| XM_021258926.1 | XP_021114585.1 | 1 - 2 | 336 | 69 | ||
More Information
Set Membership: |
|

Desi1
Genomic Map