Assay Details
| Gene Symbol: Bckdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860296.3 | XP_004860353.2 | 2 - 3 | 331 | 85 | |
| XM_013077973.2 | XP_012933427.2 | 2 - 3 | 331 | 85 | ||
| XM_021240690.1 | XP_021096349.1 | 2 - 3 | 331 | 85 | ||
| XM_021240691.1 | XP_021096350.1 | 2 - 3 | 331 | 85 | ||
| XM_021240692.1 | XP_021096351.1 | 2 - 3 | 331 | 85 | ||
| XM_021240693.1 | XP_021096352.1 | 2 - 3 | 103 | 85 | ||
| XM_021240694.1 | XP_021096353.1 | 2 - 3 | 103 | 85 | ||
More Information
Set Membership: |
|

Bckdhb
Genomic Map