Assay Details
| Gene Symbol: Gyg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834417.3 | XP_004834474.1 | 3 - 4 | 543 | 80 | |
| XM_004834418.3 | XP_004834475.1 | 3 - 4 | 543 | 80 | ||
| XM_021252941.1 | XP_021108600.1 | 3 - 4 | 379 | 80 | ||
More Information
Set Membership: |
|

Gyg1
Genomic Map