Assay Details
| Gene Symbol: CUNH12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861393.2 | XP_004861450.1 | 9 - 10 | 1546 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH12orf56| Gene Symbol: CUNH12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861393.2 | XP_004861450.1 | 9 - 10 | 1546 | 126 | |
Set Membership: |
|