Assay Details
| Gene Symbol: LOC101714047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849170.3 | XP_004849227.2 | 12 - 13 | 1371 | 59 | |
| XM_004849171.3 | XP_004849228.1 | 13 - 14 | 1705 | 59 | ||
| XM_004849174.3 | XP_004849231.1 | 12 - 13 | 1394 | 59 | ||
| XM_004849175.3 | XP_004849232.1 | 12 - 13 | 1370 | 59 | ||
| XM_013073651.2 | XP_012929105.1 | 13 - 14 | 1445 | 59 | ||
| XM_021261904.1 | XP_021117563.1 | 13 - 14 | 2539 | 59 | ||
| XM_021261905.1 | XP_021117564.1 | 14 - 15 | 1866 | 59 | ||
More Information
Set Membership: |
|

LOC101714047
Genomic Map