Assay Details
| Gene Symbol: LOC101714047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849170.3 | XP_004849227.2 | 2 - 3 | 217 | 71 | |
| XM_004849171.3 | XP_004849228.1 | 3 - 4 | 551 | 71 | ||
| XM_004849174.3 | XP_004849231.1 | 2 - 3 | 240 | 71 | ||
| XM_004849175.3 | XP_004849232.1 | 2 - 3 | 216 | 71 | ||
| XM_013073651.2 | XP_012929105.1 | 3 - 4 | 291 | 71 | ||
| XM_021261904.1 | XP_021117563.1 | 3 - 4 | 1385 | 71 | ||
| XM_021261905.1 | XP_021117564.1 | 4 - 5 | 712 | 71 | ||
More Information
Set Membership: |
|

LOC101714047
Genomic Map