Assay Details
| Gene Symbol: LOC101714047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849170.3 | XP_004849227.2 | 6 - 7 | 735 | 67 | |
| XM_004849171.3 | XP_004849228.1 | 7 - 8 | 1069 | 67 | ||
| XM_004849174.3 | XP_004849231.1 | 6 - 7 | 758 | 67 | ||
| XM_004849175.3 | XP_004849232.1 | 6 - 7 | 734 | 67 | ||
| XM_013073651.2 | XP_012929105.1 | 7 - 8 | 809 | 67 | ||
| XM_021261904.1 | XP_021117563.1 | 7 - 8 | 1903 | 67 | ||
| XM_021261905.1 | XP_021117564.1 | 8 - 9 | 1230 | 67 | ||
More Information
Set Membership: |
|

LOC101714047
Genomic Map