Assay Details
| Gene Symbol: Ntmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849138.2 | XP_004849195.1 | 3 - 4 | 718 | 80 | |
| XM_013073911.2 | XP_012929365.1 | 2 - 3 | 386 | 80 | ||
| XM_021261890.1 | XP_021117549.1 | 3 - 4 | 658 | 80 | ||
| XM_021261891.1 | XP_021117550.1 | 2 - 3 | 278 | 80 | ||
More Information
Set Membership: |
|

Ntmt1
Genomic Map