Assay Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854525.3 | XP_004854582.1 | 5 - 6 | 888 | 65 | |
| XM_013075821.2 | XP_012931275.1 | 4 - 5 | 798 | 65 | ||
More Information
Set Membership: |
|

Il15ra
Genomic Map