Assay Details
| Gene Symbol: Chrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851236.2 | XP_004851293.1 | 2 - 3 | 112 | 110 | |
| XM_021263642.1 | XP_021119301.1 | 1 - 2 | 305 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004851236.2, XM_021263642.1 transcripts.
Set Membership: |
|

Chrm2
Genomic Map