Assay Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837680.2 | XP_004837737.1 | 2 - 3 | 184 | 108 | |
| XM_021250084.1 | XP_021105743.1 | 3 - 4 | 244 | 108 | ||
More Information
Set Membership: |
|

Lrrc24
Genomic Map