Assay Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865391.3 | XP_004865448.1 | 2 - 3 | 274 | 66 | |
| XM_004865392.3 | XP_004865449.1 | 2 - 3 | 274 | 66 | ||
More Information
Set Membership: |
|

Elavl1
Genomic Map