Assay Details
| Gene Symbol: Vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839022.3 | XP_004839079.1 | 2 - 3 | 594 | 72 | |
| XM_004839023.3 | XP_004839080.1 | 2 - 3 | 474 | 72 | ||
| XM_013070072.2 | XP_012925526.1 | 2 - 3 | 346 | 72 | ||
More Information
Set Membership: |
|

Vsnl1
Genomic Map