Assay Details
| Gene Symbol: Chi3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862192.2 | XP_004862249.1 | 7 - 8 | 811 | 65 | |
| XM_021242503.1 | XP_021098162.1 | 7 - 8 | 816 | 65 | ||
More Information
Set Membership: |
|

Chi3l1
Genomic Map