Assay Details
| Gene Symbol: Cenps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863607.2 | XP_004863664.1 | 1 - 2 | 242 | 66 | |
| XM_004863614.3 | XP_004863671.1 | 1 - 2 | 204 | 66 | ||
More Information
Set Membership: |
|

Cenps
Genomic Map