Assay Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870275.3 | XP_004870332.1 | 4 - 5 | 666 | 57 | |
| XM_004870277.3 | XP_004870334.1 | 4 - 5 | 904 | 57 | ||
More Information
Set Membership: |
|

Amz2
Genomic Map