Assay Details
| Gene Symbol: Map3k8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861784.3 | XP_004861841.1 | 6 - 7 | 1281 | 67 | |
| XM_004861785.2 | XP_004861842.1 | 6 - 7 | 1163 | 67 | ||
| XM_021242038.1 | XP_021097697.1 | 6 - 7 | 1275 | 67 | ||
More Information
Set Membership: |
|

Map3k8
Genomic Map