Assay Details
| Gene Symbol: Cln8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869561.3 | XP_004869618.1 | 2 - 3 | 1846 | 99 | |
| XM_004869564.3 | XP_004869621.1 | 2 - 3 | 1548 | 99 | ||
| XM_004869565.2 | XP_004869622.1 | 2 - 3 | 772 | 99 | ||
| XM_004869566.3 | XP_004869623.1 | 2 - 3 | 2099 | 99 | ||
| XM_013067381.2 | XP_012922835.1 | 2 - 3 | 692 | 99 | ||
| XM_021248177.1 | XP_021103836.1 | 3 - 4 | 1453 | 99 | ||
| XM_021248178.1 | XP_021103837.1 | 3 - 4 | 3943 | 99 | ||
| XM_021248179.1 | XP_021103838.1 | 3 - 4 | 1974 | 99 | ||
| XM_021248180.1 | XP_021103839.1 | 3 - 4 | 3392 | 99 | ||
| XM_021248181.1 | XP_021103840.1 | 3 - 4 | 902 | 99 | ||
| XM_021248182.1 | XP_021103841.1 | 2 - 3 | 1444 | 99 | ||
More Information
Set Membership: |
|

Cln8
Genomic Map