Assay Details
| Gene Symbol: Sumf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854233.2 | XP_004854290.1 | 8 - 9 | 1026 | 67 | |
| XM_021266054.1 | XP_021121713.1 | 12 - 13 | 1383 | 67 | ||
| XM_021266055.1 | XP_021121714.1 | 12 - 13 | 1377 | 67 | ||
| XM_021266056.1 | XP_021121715.1 | 11 - 12 | 1320 | 67 | ||
| XM_021266057.1 | XP_021121716.1 | 10 - 11 | 1254 | 67 | ||
| XM_021266058.1 | XP_021121717.1 | 10 - 11 | 1245 | 67 | ||
| XM_021266059.1 | XP_021121718.1 | 11 - 12 | 1218 | 67 | ||
| XM_021266060.1 | XP_021121719.1 | 11 - 12 | 1212 | 67 | ||
| XM_021266061.1 | XP_021121720.1 | 11 - 12 | 1324 | 67 | ||
| XM_021266062.1 | XP_021121721.1 | 11 - 12 | 1318 | 67 | ||
| XM_021266064.1 | XP_021121723.1 | 10 - 11 | 1155 | 67 | ||
| XM_021266065.1 | XP_021121724.1 | 9 - 10 | 1089 | 67 | ||
| XM_021266066.1 | XP_021121725.1 | 7 - 8 | 951 | 67 | ||
More Information
Set Membership: |
|

Sumf1
Genomic Map