Assay Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873040.3 | XP_004873097.1 | 2 - 3 | 522 | 65 | |
| XM_021250983.1 | XP_021106642.1 | 2 - 3 | 526 | 65 | ||
More Information
Set Membership: |
|

Dedd2
Genomic Map