Assay Details
| Gene Symbol: CUNH16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843068.3 | XP_004843125.1 | 12 - 13 | 1118 | 114 | |
| XM_004843069.3 | XP_004843126.1 | 12 - 13 | 1150 | 114 | ||
| XM_021256904.1 | XP_021112563.1 | 11 - 12 | 1034 | 114 | ||
More Information
Set Membership: |
|

CUNH16orf70
Genomic Map