Assay Details
| Gene Symbol: CUNH16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843068.3 | XP_004843125.1 | 3 - 4 | 294 | 103 | |
| XM_004843069.3 | XP_004843126.1 | 3 - 4 | 326 | 103 | ||
More Information
Set Membership: |
|

CUNH16orf70
Genomic Map