Assay Details
| Gene Symbol: CUNH16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843068.3 | XP_004843125.1 | 8 - 9 | 700 | 103 | |
| XM_004843069.3 | XP_004843126.1 | 8 - 9 | 732 | 103 | ||
| XM_021256904.1 | XP_021112563.1 | 7 - 8 | 616 | 103 | ||
More Information
Set Membership: |
|

CUNH16orf70
Genomic Map