Assay Details
| Gene Symbol: Clybl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858621.2 | XP_004858678.1 | 3 - 4 | 480 | 70 | |
| XM_004858622.3 | XP_004858679.1 | 3 - 4 | 480 | 70 | ||
More Information
Set Membership: |
|

Clybl
Genomic Map