Assay Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856968.3 | XP_004857025.2 | 2 - 3 | 240 | 76 | |
| XM_021238124.1 | XP_021093783.1 | 2 - 3 | 265 | 76 | ||
| XM_021238125.1 | XP_021093784.1 | 3 - 4 | 413 | 76 | ||
| XM_021238126.1 | XP_021093785.1 | 2 - 3 | 162 | 76 | ||
| XM_021238127.1 | XP_021093786.1 | 2 - 3 | 224 | 76 | ||
| XM_021238129.1 | XP_021093788.1 | 2 - 3 | 453 | 76 | ||
More Information
Set Membership: |
|

Lyrm9
Genomic Map