Assay Details
| Gene Symbol: Cwc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838056.3 | XP_004838113.1 | 6 - 7 | 756 | 69 | |
| XM_021252674.1 | XP_021108333.1 | 6 - 7 | 797 | 69 | ||
More Information
Set Membership: |
|

Cwc15
Genomic Map