Assay Details
| Gene Symbol: LOC101710885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861558.2 | XP_004861615.1 | 5 - 6 | 609 | 73 | |
| XM_021241731.1 | XP_021097390.1 | 12 - 13 | 1211 | 73 | ||
| XM_021241732.1 | XP_021097391.1 | 12 - 13 | 1211 | 73 | ||
| XM_021241733.1 | XP_021097392.1 | 12 - 13 | 1211 | 73 | ||
More Information
Set Membership: |
|

LOC101710885
Genomic Map