Assay Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836892.2 | XP_004836949.1 | 3 - 4 | 446 | 65 | |
| XM_021243865.1 | XP_021099524.1 | 3 - 4 | 410 | 65 | ||
More Information
Set Membership: |
|

Ckb
Genomic Map