Assay Details
| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853141.3 | XP_004853198.1 | 9 - 10 | 846 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impact| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853141.3 | XP_004853198.1 | 9 - 10 | 846 | 64 | |
Set Membership: |
|