Assay Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074953.2 | XP_012930407.1 | 4 - 5 | 904 | 58 | |
XM_021264549.1 | XP_021120208.1 | 5 - 6 | 1038 | 58 | ||
XM_021264551.1 | XP_021120210.1 | 5 - 6 | 886 | 58 | ||
XM_021264552.1 | XP_021120211.1 | 4 - 5 | 599 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
