Assay Details
| Gene Symbol: Sptssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253826.1 | XP_021109485.1 | 2 - 3 | 1724 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021253826.1 transcripts.
Set Membership: |
|

Sptssb
Genomic Map